Learn More
Selleck Chemical LLC Catalpol S3909-25mg
SDP
Catalog No. 503111482
Encompass_Preferred
Catalpol (Catalpinoside Digitalis purpurea L) an iridoid glucoside contained richly in the roots of the small flowering plant species Rehmannia glutinosa Libosch has been shown to have antioxidation anti-inflammation anti-apoptosis and other neuroprotective properties and plays a role in neuroprotection against hypoxic/ischemic injury AD and PD in both in vivo and in vitro models
Catalog No. 50-311-1482 Supplier Selleck Chemical LLC Supplier No. S390925MG
Catalpol (Catalpinoside Digitalis purpurea L) an iridoid glucoside contained richly in the roots of the small flowering plant species Rehmannia glutinosa Libosch has been shown to have antioxidation anti-inflammation anti-apoptosis and other neuroprotective properties and plays a role in neuroprotection against hypoxic/ischemic injury AD and PD in both in vivo and in vitro models