Learn More
Medchemexpress LLC Catalposide | 6736-85-2 | 5 MG
SDP
Catalog No. 5000418425
Encompass_Preferred
Catalposide is an iridoid glycoside isolated from Catalpa ovata G. Don (Bignoniaceae). It inhibits the production of TNF-α, IL-1β, and IL-6, and also suppresses NF-κB (p65) activation in lipopolysaccharide-activated RAW 264.7 macrophages. It is intended for research use only.
  • Purity of 98.0%
  • Inhibits TNF-α, IL-1β, and IL-6 production
  • Suppresses NF-κB (p65) activation in lipopolysaccharide-activated RAW 264.7 macrophages
  • Displays IC50 > 10 μM against human A2780, A549, BGC-823, Bel-7402, and HT-29 cells
  • Isolated from Catalpa ovata G. Don (Bignoniaceae)
Catalog No. 50-004-18425 Supplier Medchemexpress LLC Supplier No. HYN35525MG
Catalposide is an iridoid glycoside isolated from Catalpa ovata G. Don (Bignoniaceae). It inhibits the production of TNF-α, IL-1β, and IL-6, and also suppresses NF-κB (p65) activation in lipopolysaccharide-activated RAW 264.7 macrophages. It is intended for research use only.
  • Purity of 98.0%
  • Inhibits TNF-α, IL-1β, and IL-6 production
  • Suppresses NF-κB (p65) activation in lipopolysaccharide-activated RAW 264.7 macrophages
  • Displays IC50 > 10 μM against human A2780, A549, BGC-823, Bel-7402, and HT-29 cells
  • Isolated from Catalpa ovata G. Don (Bignoniaceae)