Learn More
Medchemexpress LLC Catalposide | 6736-85-2 | MFCD28015867 | 1 MG
SDP
Catalog No. 5000419036
Encompass_Preferred
Catalposide is an iridoid glycoside isolated from Catalpa ovata G. Don (Bignoniaceae). It inhibits TNF-α, IL-1β, and IL-6 productions, and NF-κB (p65) activation in lipopolysaccharide-activated RAW 264.7 macrophages. This product is for research use only.
  • Solid appearance
  • White to yellow color
  • Store at 4°C, protect from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protect from light
Catalog No. 50-004-19036 Supplier Medchemexpress LLC Supplier No. HYN35521MG
Catalposide is an iridoid glycoside isolated from Catalpa ovata G. Don (Bignoniaceae). It inhibits TNF-α, IL-1β, and IL-6 productions, and NF-κB (p65) activation in lipopolysaccharide-activated RAW 264.7 macrophages. This product is for research use only.
  • Solid appearance
  • White to yellow color
  • Store at 4°C, protect from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protect from light