Learn More
Vector Laboratories TurboLINK™ Catalyst Buffer (10X) 1.5 mL
SDP
Catalog No. 502045753
Encompass
TurboLINK Catalyst Buffer is used to catalyze the SoluLINK bioconjugation reactions, which are based on aromatic hydrazone formation between HyNic-modified biomolecules and 4FB-modified biomolecules. 2° - 8°C – Do Not Freeze
Catalog No. 50-204-5753 Supplier Vector Laboratories Supplier No. S2006105
TurboLINK Catalyst Buffer is used to catalyze the SoluLINK bioconjugation reactions, which are based on aromatic hydrazone formation between HyNic-modified biomolecules and 4FB-modified biomolecules. 2° - 8°C – Do Not Freeze