Learn More
AVANTOR MASTERFLEX (VWR LLC) Catalyst FH100, FH100D Pump Tubing, PharMed BPT, 6.4 mm ID, 1.6 mm wall, 15 m
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9700024
Encompass
Catalyst pump tubing comes in four formulations to cover most chemical compatible applications. FH100 and FH100D utilize tubing with a wall thickness of 1.6 mm. Standard 15 meter lengths.
Catalog No. NC1230538