Learn More
AVANTOR MASTERFLEX (VWR LLC) Stopped Tubing Assembly, Platinum-Cured Silicone, 1.6 mm ID, 6 pk
Catalog No. NC1536604
Encompass
Catalyst Stopped Tubing Assembly, Platinum-Cured Silicone, 1.6 mm ID, 6 pk, Start pumping with little effort - preconfigured Links for quick and easy setupFour formulations offer wide range of chemical compatibility.Save time with already assembled links.Use with the Catalyst extension coils / tubing to complete your system.FH15 and FH30 utilize the stopped assemblies that have an ID of: 0.8, 1.6, 3.2, and 4.8 mm.
Catalog No. NC1536604 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9710013
Catalyst Stopped Tubing Assembly, Platinum-Cured Silicone, 1.6 mm ID, 6 pk, Start pumping with little effort - preconfigured Links for quick and easy setupFour formulations offer wide range of chemical compatibility.Save time with already assembled links.Use with the Catalyst extension coils / tubing to complete your system.FH15 and FH30 utilize the stopped assemblies that have an ID of: 0.8, 1.6, 3.2, and 4.8 mm.