Learn More
Med Vet International Catch-A-Caca, J1525, Each
Catalog No. 5000495914
Encompass_Preferred
Cleaning up after pets is an everyday necessity at a vet clinic. The Catch-A-Caca is a useful device that can catch fecal material in a sanitary bag as the dog defecates while on a walk outside. This makes clean up faster and cleaner, especially with loose stool or diarrhea cases. • Uses standard "poop" bags that attach easily to distal end • Telescoping shaft reaches up to 3 ft. • Super light weight aluminum, just 9 oz. • Lanyard to hold securely • Soft grip • Collapses to easily hang on belt or shirt pocket • Comes with 15 bags Replacement bags use the Ecobag replacement roll (J1102b)
Catalog No. 50-004-95914 Supplier Med Vet International Supplier No. JOR1525
Cleaning up after pets is an everyday necessity at a vet clinic. The Catch-A-Caca is a useful device that can catch fecal material in a sanitary bag as the dog defecates while on a walk outside. This makes clean up faster and cleaner, especially with loose stool or diarrhea cases. • Uses standard "poop" bags that attach easily to distal end • Telescoping shaft reaches up to 3 ft. • Super light weight aluminum, just 9 oz. • Lanyard to hold securely • Soft grip • Collapses to easily hang on belt or shirt pocket • Comes with 15 bags Replacement bags use the Ecobag replacement roll (J1102b)