Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Devices Molecular Devices cAMP Fluorescent Assay Kit, 384-Well, Bulk, 60 μL

Catalog No. 5000014448
Encompass_Preferred

The CatchPoint cAMP Fluorescent Assay Kit is a high-throughput competitive immunoassay for quantifying 3',5'-cyclic adenosine monophosphate (cAMP) and measuring adenylate cyclase activity in biochemical and cell-based assays. The 384-well bulk format supplies reagents for 20 plates (7,680 assay points) and is optimized for fluorescence intensity detection with long signal stability.

  • 384-well bulk format, sufficient for 20 plates (7,680 wells)
  • Reaction volume 60 μL per well, 50 μL supported for some HTS systems
  • Fluorescence intensity detection using Stoplight Red substrate
  • Minimum detectable concentration 0.1 nM (6 fmol/well), EC50 approximately 3.4 nM
  • Readings stable from 10 minutes up to 24 hours after substrate addition
  • Store components at 4°C; kit components stable for six months when stored properly

Catalog No. 50-000-14448 Supplier Molecular Devices Supplier No. R8053
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The CatchPoint cAMP Fluorescent Assay Kit is a high-throughput competitive immunoassay for quantifying 3',5'-cyclic adenosine monophosphate (cAMP) and measuring adenylate cyclase activity in biochemical and cell-based assays. The 384-well bulk format supplies reagents for 20 plates (7,680 assay points) and is optimized for fluorescence intensity detection with long signal stability.

  • 384-well bulk format, sufficient for 20 plates (7,680 wells)
  • Reaction volume 60 μL per well, 50 μL supported for some HTS systems
  • Fluorescence intensity detection using Stoplight Red substrate
  • Minimum detectable concentration 0.1 nM (6 fmol/well), EC50 approximately 3.4 nM
  • Readings stable from 10 minutes up to 24 hours after substrate addition
  • Store components at 4°C; kit components stable for six months when stored properly

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.