Learn More
Molecular Devices Molecular Devices CatchPoint cGMP Assay Kit, 384-Well, Clear Bottom
Catalog No. 5000014437
Encompass_Preferred
The CatchPoint cGMP Fluorescent Assay Kit Explorer configuration is a high-sensitivity, cell-compatible immunoassay for quantitative measurement of cyclic GMP in high-throughput formats. The kit provides pre-coated 384-well plates and optimized reagents for detecting low cGMP levels with a stable fluorescent readout and flexible timing for automation.
  • Pre-coated, pre-blocked 384-well black well, clear bottom plates, two plates per kit
  • Includes reagents and buffers for assay setup and detection, including antibody, conjugate, calibrator, and substrate
  • Fluorescent Stoplight Red readout with stable signal from 10 minutes up to 24 hours
  • Quantitative competitive immunoassay format suitable for cell-based assays and HTS
  • Robust format resists interference from colored or fluorescent test compounds
Catalog No. 50-000-14437 Supplier Molecular Devices Supplier No. R8065
The CatchPoint cGMP Fluorescent Assay Kit Explorer configuration is a high-sensitivity, cell-compatible immunoassay for quantitative measurement of cyclic GMP in high-throughput formats. The kit provides pre-coated 384-well plates and optimized reagents for detecting low cGMP levels with a stable fluorescent readout and flexible timing for automation.
  • Pre-coated, pre-blocked 384-well black well, clear bottom plates, two plates per kit
  • Includes reagents and buffers for assay setup and detection, including antibody, conjugate, calibrator, and substrate
  • Fluorescent Stoplight Red readout with stable signal from 10 minutes up to 24 hours
  • Quantitative competitive immunoassay format suitable for cell-based assays and HTS
  • Robust format resists interference from colored or fluorescent test compounds