Learn More
Molecular Devices Molecular Devices Membrane Potential Assay Kit, Red, Explorer, Homogeneous Fluorescence
Catalog No. 5000014340
Encompass_Preferred
The FLIPR Membrane Potential Assay Kit (Red, Explorer) is a homogeneous, fluorescence-based reagent kit for sensitive detection of rapid changes in cellular membrane potential. It is optimized for high-throughput screening and fast kinetic measurements and is compatible with automated plate readers and FLIPR/Tetra systems.
  • Homogeneous fluorescence formulation for rapid membrane potential detection.
  • Red indicator optimized for larger signal window and temperature stability.
  • Explorer format supplies reagents for approximately 10-15 microplates.
  • Compatible with 96-, 384-, and 1536-well plate formats.
  • Suitable for ion channel and transporter assays, including TRP and voltage-gated channels.
  • Free of animal-derived components.
Catalog No. 50-000-14340 Supplier Molecular Devices Supplier No. R8126
The FLIPR Membrane Potential Assay Kit (Red, Explorer) is a homogeneous, fluorescence-based reagent kit for sensitive detection of rapid changes in cellular membrane potential. It is optimized for high-throughput screening and fast kinetic measurements and is compatible with automated plate readers and FLIPR/Tetra systems.
  • Homogeneous fluorescence formulation for rapid membrane potential detection.
  • Red indicator optimized for larger signal window and temperature stability.
  • Explorer format supplies reagents for approximately 10-15 microplates.
  • Compatible with 96-, 384-, and 1536-well plate formats.
  • Suitable for ion channel and transporter assays, including TRP and voltage-gated channels.
  • Free of animal-derived components.