Learn More
Molecular Devices Molecular Devices Fluorescent Assay Kit, cGMP, 96-Well, Competitive Immunoassay
Catalog No. 5000014481
Encompass_Preferred
The CatchPoint cGMP Fluorescent Assay Kit is a 96-well competitive immunoassay for quantitative measurement of 3',5'-cyclic guanosine monophosphate (cGMP) and guanylate cyclase activity in biochemical and cell-based assays. The kit provides sensitive fluorescence-based detection and includes pre-coated microplates and optimized reagents for high-throughput workflows.
  • 96-well format with a black microplate and clear bottom.
  • Includes reagents and buffers for assay setup and detection.
  • Reagents sufficient for two 96-well plates (192 reactions at 120 μL).
  • Fluorescence detection using Stoplight Red substrate, compatible with common plate readers.
  • Typical sensitivity down to ~0.2 nM cGMP, dynamic range up to ~120 nM.
  • Store kit components at 4°C; reconstituted working solutions stable for about three weeks at 4°C.
Catalog No. 50-000-14481 Supplier Molecular Devices Supplier No. R8074
The CatchPoint cGMP Fluorescent Assay Kit is a 96-well competitive immunoassay for quantitative measurement of 3',5'-cyclic guanosine monophosphate (cGMP) and guanylate cyclase activity in biochemical and cell-based assays. The kit provides sensitive fluorescence-based detection and includes pre-coated microplates and optimized reagents for high-throughput workflows.
  • 96-well format with a black microplate and clear bottom.
  • Includes reagents and buffers for assay setup and detection.
  • Reagents sufficient for two 96-well plates (192 reactions at 120 μL).
  • Fluorescence detection using Stoplight Red substrate, compatible with common plate readers.
  • Typical sensitivity down to ~0.2 nM cGMP, dynamic range up to ~120 nM.
  • Store kit components at 4°C; reconstituted working solutions stable for about three weeks at 4°C.