Learn More
Molecular Devices Molecular Devices CatchPoint cAMP Assay Kit, 96-Well, Fluorescent, Explorer
Catalog No. 5000014436
Encompass_Preferred
The CatchPoint cAMP Fluorescent Assay Kit in 96-well Explorer format provides reagents and pre-coated plates for sensitive fluorescence-based measurement of cyclic AMP and adenylate cyclase activity in cell-based and biochemical assays.
  • High-sensitivity fluorescent competitive immunoassay for cAMP quantification.
  • Includes two pre-coated 96-well black well/clear bottom plates and reagents for 192 reactions.
  • Pre-blocked plates reduce background and simplify assay setup.
  • Complete reagent set and buffers for an ELISA-style workflow, including antibody, calibrator, conjugate, and substrate.
  • Compatible with standard fluorescence microplate readers and high-throughput screening systems.
Catalog No. 50-000-14436 Supplier Molecular Devices Supplier No. R8088
The CatchPoint cAMP Fluorescent Assay Kit in 96-well Explorer format provides reagents and pre-coated plates for sensitive fluorescence-based measurement of cyclic AMP and adenylate cyclase activity in cell-based and biochemical assays.
  • High-sensitivity fluorescent competitive immunoassay for cAMP quantification.
  • Includes two pre-coated 96-well black well/clear bottom plates and reagents for 192 reactions.
  • Pre-blocked plates reduce background and simplify assay setup.
  • Complete reagent set and buffers for an ELISA-style workflow, including antibody, calibrator, conjugate, and substrate.
  • Compatible with standard fluorescence microplate readers and high-throughput screening systems.