Learn More
Molecular Devices Molecular Devices cGMP Fluorescent Assay Kit, 96-Well, Bulk, Competitive Immunoassay
Catalog No. 5000014456
Encompass_Preferred
CatchPoint cGMP Fluorescent Assay Bulk Kit provides a high-sensitivity, 96-well competitive immunoassay for quantitative measurement of cGMP and guanylate cyclase activity. The bulk format supplies reagents and ten pre-coated plates for high-throughput biochemical and cell-based assays.
  • High-affinity reagents optimized for low cGMP levels.
  • Ten pre-coated black clear-bottom 96-well plates in bulk format.
  • Sufficient reagents for ten 96-well plates (960 reactions at 120 μL per well).
  • Competitive immunoassay with a single washing step.
  • Flexible read window: 10 minutes to 24 hours after substrate addition.
  • Store components at 4°C; unopened kit stable for six months.
Catalog No. 50-000-14456 Supplier Molecular Devices Supplier No. R8075
Add to Cart
Add to Cart
Request a Quote
CatchPoint cGMP Fluorescent Assay Bulk Kit provides a high-sensitivity, 96-well competitive immunoassay for quantitative measurement of cGMP and guanylate cyclase activity. The bulk format supplies reagents and ten pre-coated plates for high-throughput biochemical and cell-based assays.
  • High-affinity reagents optimized for low cGMP levels.
  • Ten pre-coated black clear-bottom 96-well plates in bulk format.
  • Sufficient reagents for ten 96-well plates (960 reactions at 120 μL per well).
  • Competitive immunoassay with a single washing step.
  • Flexible read window: 10 minutes to 24 hours after substrate addition.
  • Store components at 4°C; unopened kit stable for six months.