Learn More
Medchemexpress LLC HY-N0898 10mg Medchemexpress, Catechin CAS:154-23-4 Purity:>98%
SDP
Catalog No. 501933783
Encompass_Preferred
Medchemexpress, HY-N0898 10mg Catechin CAS:154-23-4 Catechin inhibits cyclooxygenase-1 ( COX-1 ) with an IC 50 of 1.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3783 Supplier Medchemexpress LLC Supplier No. HYN089810MG
Medchemexpress, HY-N0898 10mg Catechin CAS:154-23-4 Catechin inhibits cyclooxygenase-1 ( COX-1 ) with an IC 50 of 1.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.