Learn More
Medchemexpress LLC Catechin-13C3 | 154-23-4 | MFCD15144883 | 293.25 | 1 MG
SDP
Catalog No. 5000416467
Encompass_Preferred
Catechin-13C3 is a 13C-labeled version of Catechin, a natural antioxidant found in various plants. This compound inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM. This stable isotope-labeled compound is valuable for research applications.
  • Labeled stable isotope
  • Useful for metabolic studies
  • Ideal for quantitative analysis
  • Suitable for tracking biochemical pathways
Catalog No. 50-004-16467 Supplier Medchemexpress LLC Supplier No. HYN0898S1MG
Catechin-13C3 is a 13C-labeled version of Catechin, a natural antioxidant found in various plants. This compound inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM. This stable isotope-labeled compound is valuable for research applications.
  • Labeled stable isotope
  • Useful for metabolic studies
  • Ideal for quantitative analysis
  • Suitable for tracking biochemical pathways