Learn More
Apexbio Technology LLC Catechin 154-23-4 100mg
SDP
Catalog No. 5000094913
Encompass_Preferred
Catechin (154-23-4) is a small-molecule inhibitor targeting cyclooxygenase-1 (COX-1) It is designed to inhibit COX-1 activity thereby modulating inflammation-related pathways Catechin exerts its biological activity primarily through inhibition of COX-1 In cell-based assays catechin demonstrates inhibitory activity with an IC50 value of approximately 1 4 M Based on these pharmacological properties catechin holds research potential in studies of oxidative stress pathways inflammation processes and enzyme inhibition screening
Catalog No. 50-000-94913 Supplier Apexbio Technology LLC Supplier No. A4629100
Catechin (154-23-4) is a small-molecule inhibitor targeting cyclooxygenase-1 (COX-1) It is designed to inhibit COX-1 activity thereby modulating inflammation-related pathways Catechin exerts its biological activity primarily through inhibition of COX-1 In cell-based assays catechin demonstrates inhibitory activity with an IC50 value of approximately 1 4 M Based on these pharmacological properties catechin holds research potential in studies of oxidative stress pathways inflammation processes and enzyme inhibition screening