Learn More
Apexbio Technology LLC Catechin 154-23-4 10mg
SDP
Catalog No. 5000093680
Encompass_Preferred
Catechin (154-23-4) is a small-molecule inhibitor targeting cyclooxygenase-1 (COX-1) It is designed to inhibit COX-1 activity thereby modulating inflammation-related pathways Catechin exerts its biological activity primarily through inhibition of COX-1 In cell-based assays catechin demonstrates inhibitory activity with an IC50 value of approximately 1 4 M Based on these pharmacological properties catechin holds research potential in studies of oxidative stress pathways inflammation processes and enzyme inhibition screening
Catalog No. 50-000-93680 Supplier Apexbio Technology LLC Supplier No. A462910
Catechin (154-23-4) is a small-molecule inhibitor targeting cyclooxygenase-1 (COX-1) It is designed to inhibit COX-1 activity thereby modulating inflammation-related pathways Catechin exerts its biological activity primarily through inhibition of COX-1 In cell-based assays catechin demonstrates inhibitory activity with an IC50 value of approximately 1 4 M Based on these pharmacological properties catechin holds research potential in studies of oxidative stress pathways inflammation processes and enzyme inhibition screening