Learn More
Medchemexpress LLC Catechin | 154-23-4 | 290.27 | 10 MM 1 ML
SDP
Catalog No. 5000416888
Encompass_Preferred
Catechin ((+)-Catechin) inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM. It exhibits potent inhibitory activity against COX-1 and can induce apoptosis in certain cancer cells. Catechin has also shown a dose-dependent reduction in cell viability and an increase in apoptosis markers in vitro. In animal studies, it has demonstrated the ability to prevent episodic memory deficits.
  • Inhibits cyclooxygenase-1.
  • Reduces cell viability in a dose-dependent manner.
  • Increases the induction of apoptosis.
  • Prevents time-induced episodic memory deficits.
Catalog No. 50-004-16888 Supplier Medchemexpress LLC Supplier No. HYN089810MM/1ML
Catechin ((+)-Catechin) inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM. It exhibits potent inhibitory activity against COX-1 and can induce apoptosis in certain cancer cells. Catechin has also shown a dose-dependent reduction in cell viability and an increase in apoptosis markers in vitro. In animal studies, it has demonstrated the ability to prevent episodic memory deficits.
  • Inhibits cyclooxygenase-1.
  • Reduces cell viability in a dose-dependent manner.
  • Increases the induction of apoptosis.
  • Prevents time-induced episodic memory deficits.