Learn More
Medchemexpress LLC Catechin | 154-23-4 | 290.27 | 100 MG
SDP
Catalog No. 5000416466
Encompass_Preferred
Catechin ((+)-Catechin) inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM.
  • Mechanism of action: inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM and exhibits >95% inhibitory activity at 70 μg/mL against COX-1.
  • Cellular effects: induces apoptosis in MCF-7 cells in a dose- and time-dependent manner, leading to increased expression levels of Caspase-3, -8, -9, and p53.
  • In vivo effects: prevents time-induced episodic memory deficits in animals in a dose-dependent manner (most effective at 200 mg/kg, p.o.) and prevents the rise in MPO level.
Catalog No. 50-004-16466 Supplier Medchemexpress LLC Supplier No. HYN0898100MG
Catechin ((+)-Catechin) inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM.
  • Mechanism of action: inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM and exhibits >95% inhibitory activity at 70 μg/mL against COX-1.
  • Cellular effects: induces apoptosis in MCF-7 cells in a dose- and time-dependent manner, leading to increased expression levels of Caspase-3, -8, -9, and p53.
  • In vivo effects: prevents time-induced episodic memory deficits in animals in a dose-dependent manner (most effective at 200 mg/kg, p.o.) and prevents the rise in MPO level.