Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Catechin | 154-23-4 | 290.27 | 5 MG
SDP

Catalog No. 5000416071
Encompass_Preferred

Catechin ((+)-Catechin) inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM.

  • 15+ Cited publications
  • Also known as (+)-Catechin; Cianidanol; Catechuic acid
  • Targeting COX apoptosis, influenza virus, and endogenous metabolite pathways
  • Research area: Cancer
  • Structural classification: Anti-aging, Camellia sinensis (L.) O. Ktze. plants, endogenous metabolite, flavonoids, sunscreen, flavanols, phenols, polyphenols
  • Application fields: Cosmetic research, disease research fields
  • Source classification: Theaceae

Catalog No. 50-004-16071 Supplier Medchemexpress LLC Supplier No. HYN08985MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Catechin ((+)-Catechin) inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM.

  • 15+ Cited publications
  • Also known as (+)-Catechin; Cianidanol; Catechuic acid
  • Targeting COX apoptosis, influenza virus, and endogenous metabolite pathways
  • Research area: Cancer
  • Structural classification: Anti-aging, Camellia sinensis (L.) O. Ktze. plants, endogenous metabolite, flavonoids, sunscreen, flavanols, phenols, polyphenols
  • Application fields: Cosmetic research, disease research fields
  • Source classification: Theaceae

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.