Learn More
Medchemexpress LLC Catechin | 154-23-4 | MFCD00075649 | 290.27 | 50 MG
SDP
Catalog No. 5000417023
Encompass_Preferred
Catechin, also known as (+)-Catechin, acts as an inhibitor for cyclooxygenase-1 (COX-1). It exhibits an IC50 value of 1.4 μM, highlighting its activity against this enzyme.
  • Inhibits cyclooxygenase-1 (COX-1)
  • Features an IC50 of 1.4 μM
Catalog No. 50-004-17023 Supplier Medchemexpress LLC Supplier No. HYN089850MG
Catechin, also known as (+)-Catechin, acts as an inhibitor for cyclooxygenase-1 (COX-1). It exhibits an IC50 value of 1.4 μM, highlighting its activity against this enzyme.
  • Inhibits cyclooxygenase-1 (COX-1)
  • Features an IC50 of 1.4 μM