Learn More
Medchemexpress LLC (-)-Catechin | 18829-70-4 | 290.27 | 1 ML
SDP
Catalog No. 5000416469
Encompass_Preferred
(-)-Catechin is a structural isomer of Catechin. It inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM. Additionally, (-)-Catechin promotes the differentiation of hBM-MSC adipose cells, leading to increased fat cell differentiation and PPARγ levels. It is intended for research use only.
  • Inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM.
  • Promotes adipocyte differentiation in human bone marrow mesenchymal stem cells (hBM-MSC).
  • Increases fat cell differentiation.
  • Increases PPARγ level.
Catalog No. 50-004-16469 Supplier Medchemexpress LLC Supplier No. HYN0898A10MM/1ML
(-)-Catechin is a structural isomer of Catechin. It inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM. Additionally, (-)-Catechin promotes the differentiation of hBM-MSC adipose cells, leading to increased fat cell differentiation and PPARγ levels. It is intended for research use only.
  • Inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM.
  • Promotes adipocyte differentiation in human bone marrow mesenchymal stem cells (hBM-MSC).
  • Increases fat cell differentiation.
  • Increases PPARγ level.