Learn More
Medchemexpress LLC (-)-Catechin | 18829-70-4 | 290.27 | 100 MG
SDP
Catalog No. 5000416468
Encompass_Preferred
(-)-Catechin is a form of catechin that inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM. It promotes the differentiation of human bone marrow mesenchymal stem cells (hBM-MSC) into adipose cells, leading to increased fat cell differentiation and higher levels of PPARγ. This compound is intended for research use only.
  • Inhibits cyclooxygenase-1 (COX-1).
  • Promotes hBM-MSC adipose cell differentiation.
  • Increases fat cell differentiation.
  • Elevates PPARγ levels.
Catalog No. 50-004-16468 Supplier Medchemexpress LLC Supplier No. HYN0898A100MG
(-)-Catechin is a form of catechin that inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM. It promotes the differentiation of human bone marrow mesenchymal stem cells (hBM-MSC) into adipose cells, leading to increased fat cell differentiation and higher levels of PPARγ. This compound is intended for research use only.
  • Inhibits cyclooxygenase-1 (COX-1).
  • Promotes hBM-MSC adipose cell differentiation.
  • Increases fat cell differentiation.
  • Elevates PPARγ levels.