Learn More
Medchemexpress LLC (-)-Catechin | 18829-70-4 | 290.27 | 50 MG
SDP
Catalog No. 5000416916
Encompass_Preferred
(-)-Catechin is a structural variation of Catechin. It acts as an inhibitor of enzyme-1 (COX-1) with an IC50 of 1.4 μM. Additionally, (-)-Catechin promotes the differentiation of hBM-MSC adipose cells, leading to increased fat cell differentiation and elevated PPARγ levels.
  • Catechin's one kind of different structure.
  • Inhibits COX-1 with an IC50 of 1.4 μM.
  • Promotes hBM-MSC adipose cell differentiation.
  • Increases fat cell differentiation.
  • Increases PPARγ level.
Catalog No. 50-004-16916 Supplier Medchemexpress LLC Supplier No. HYN0898A50MG
(-)-Catechin is a structural variation of Catechin. It acts as an inhibitor of enzyme-1 (COX-1) with an IC50 of 1.4 μM. Additionally, (-)-Catechin promotes the differentiation of hBM-MSC adipose cells, leading to increased fat cell differentiation and elevated PPARγ levels.
  • Catechin's one kind of different structure.
  • Inhibits COX-1 with an IC50 of 1.4 μM.
  • Promotes hBM-MSC adipose cell differentiation.
  • Increases fat cell differentiation.
  • Increases PPARγ level.