Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Catechin 3-O-α-L-rhamnopyranoside | 103630-03-1 | 1 MG
SDP

Catalog No. 5000418093
Encompass_Preferred

Catechin 3-O-α-L-rhamnopyranoside is a natural product isolated from the twigs of Artocarpus lakoocha. It is intended for research use only.

  • Purity of 97.0%
  • Solid appearance, light yellow to yellow color
  • Molecular weight: 436.41
  • Chemical formula: C21H24O10
  • Optimal storage at -20°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Available documentation includes data sheet, SDS, COA, and handling instructions

Catalog No. 50-004-18093 Supplier Medchemexpress LLC Supplier No. HYN35531MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Catechin 3-O-α-L-rhamnopyranoside is a natural product isolated from the twigs of Artocarpus lakoocha. It is intended for research use only.

  • Purity of 97.0%
  • Solid appearance, light yellow to yellow color
  • Molecular weight: 436.41
  • Chemical formula: C21H24O10
  • Optimal storage at -20°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Available documentation includes data sheet, SDS, COA, and handling instructions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.