Learn More
TARGETMOL CHEMICALS INC Catechin 500MG
SDP
Catalog No. 502880769
Encompass_Preferred
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Catechin (Cianidanol) is a flavan-3-ol, a type of natural phenol and antioxidant. Purity 99.05%
Catalog No. 50-288-0769 Supplier TARGETMOL CHEMICALS INC Supplier No. T0822500MG
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Catechin (Cianidanol) is a flavan-3-ol, a type of natural phenol and antioxidant. Purity 99.05%