Learn More
Medchemexpress LLC HY-N0898A 5mg Medchemexpress, (-)-Catechin CAS:18829-70-4 Purity:>98%
SDP
Catalog No. 502031414
Encompass_Preferred
Medchemexpress, HY-N0898A 5mg (-)-Catechin CAS:18829-70-4 (-)-Catechin, isolated from green tea, is an isomer of Catechin having a trans 2S,3R configuration at the chiral center. Catechin inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1414 Supplier Medchemexpress LLC Supplier No. HYN0898A5MG
Medchemexpress, HY-N0898A 5mg (-)-Catechin CAS:18829-70-4 (-)-Catechin, isolated from green tea, is an isomer of Catechin having a trans 2S,3R configuration at the chiral center. Catechin inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.