Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Catechin 7-O-β-d-glucopyranoside | 65597-47-9 | 99.5% | 452.41 g/mol | C21H24O11 | 10 MG
SDP

Catalog No. 5000295920
Encompass_Preferred

Catechin 7-O-β-D-glucopyranoside is a flavonoid glycoside used in research for its antioxidant and anti-inflammatory properties. Supplied as a white to off-white solid with high reported purity, it is suitable for in vitro and in vivo studies and has documented uses at 10 μg/mL (in vitro) and 10 mg/kg (in vivo).

  • Appearance: White to off-white solid.
  • Purity: 99.5% (reported).
  • Molecular weight: 452.41 g/mol.
  • Solubility: DMSO 125 mg/mL (requires ultrasonic).
  • Storage: 4°C, protect from light; in solution -80°C (6 months) or -20°C (1 month).
  • Typical uses: Antioxidant and anti-inflammatory assays in cells and animal models.

Catalog No. 50-002-95920 Supplier Medchemexpress LLC Supplier No. HYN1058710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Catechin 7-O-β-D-glucopyranoside is a flavonoid glycoside used in research for its antioxidant and anti-inflammatory properties. Supplied as a white to off-white solid with high reported purity, it is suitable for in vitro and in vivo studies and has documented uses at 10 μg/mL (in vitro) and 10 mg/kg (in vivo).

  • Appearance: White to off-white solid.
  • Purity: 99.5% (reported).
  • Molecular weight: 452.41 g/mol.
  • Solubility: DMSO 125 mg/mL (requires ultrasonic).
  • Storage: 4°C, protect from light; in solution -80°C (6 months) or -20°C (1 month).
  • Typical uses: Antioxidant and anti-inflammatory assays in cells and animal models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.