Learn More
Medchemexpress LLC (±)-Catechin | 7295-85-4 | 99.5% | 1 G
SDP
Catalog No. 5000412185
Encompass_Preferred
(±)-Catechin (rel-Cianidanol) is the racemate of the green tea polyphenol Catechin. Catechin has anticancer activity and induces apoptosis. It has two forms: (+)-Catechin and its enantiomer (-)-Catechin. (+)-Catechin inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM, while (-)-Catechin promotes hBM-MSC adipocyte differentiation.
  • Exhibits anti-tumor effects
  • Shows anti-obesity and anti-diabetic properties
  • Provides anti-cardiovascular and anti-infectious benefits
  • Offers hepatoprotective and neuroprotective effects
  • Induces adipocyte differentiation
  • Increases adiponectin and PPARγ levels
Catalog No. 50-004-12185 Supplier Medchemexpress LLC Supplier No. HYB18901G
(±)-Catechin (rel-Cianidanol) is the racemate of the green tea polyphenol Catechin. Catechin has anticancer activity and induces apoptosis. It has two forms: (+)-Catechin and its enantiomer (-)-Catechin. (+)-Catechin inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM, while (-)-Catechin promotes hBM-MSC adipocyte differentiation.
  • Exhibits anti-tumor effects
  • Shows anti-obesity and anti-diabetic properties
  • Provides anti-cardiovascular and anti-infectious benefits
  • Offers hepatoprotective and neuroprotective effects
  • Induces adipocyte differentiation
  • Increases adiponectin and PPARγ levels