Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (±)-Catechin | 7295-85-4 | 99.5% | 100 MG
SDP

Catalog No. 5000409792
Encompass_Preferred

(±)-Catechin (rel-Cianidanol) is the racemate of the green tea polyphenol Catechin. Catechin has anticancer activity and induces apoptosis. It has two forms, (+)-Catechin and its enantiomer (-)-Catechin, each with distinct biological activities. The racemate exhibits anti-tumor, anti-obesity, anti-diabetic, anti-cardiovascular, anti-infectious, hepatoprotective, and neuroprotective effects.

  • Racemate of green tea polyphenol Catechin.
  • Has anticancer activity and induces apoptosis.
  • (+)-Catechin form inhibits cyclooxygenase-1 (COX-1).
  • (-)-Catechin form promotes hBM-MSC adipocyte differentiation.
  • Possesses anti-tumor, anti-obesity, anti-diabetic, anti-cardiovascular, anti-infectious, hepatoprotective, and neuroprotective effects.
  • Appearance: Solid, White to off-white.
  • Solubility: In Vitro: DMSO : 100 mg/mL.

Catalog No. 50-004-09792 Supplier Medchemexpress LLC Supplier No. HYB1890100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(±)-Catechin (rel-Cianidanol) is the racemate of the green tea polyphenol Catechin. Catechin has anticancer activity and induces apoptosis. It has two forms, (+)-Catechin and its enantiomer (-)-Catechin, each with distinct biological activities. The racemate exhibits anti-tumor, anti-obesity, anti-diabetic, anti-cardiovascular, anti-infectious, hepatoprotective, and neuroprotective effects.

  • Racemate of green tea polyphenol Catechin.
  • Has anticancer activity and induces apoptosis.
  • (+)-Catechin form inhibits cyclooxygenase-1 (COX-1).
  • (-)-Catechin form promotes hBM-MSC adipocyte differentiation.
  • Possesses anti-tumor, anti-obesity, anti-diabetic, anti-cardiovascular, anti-infectious, hepatoprotective, and neuroprotective effects.
  • Appearance: Solid, White to off-white.
  • Solubility: In Vitro: DMSO : 100 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.