Learn More
Medchemexpress LLC Catechin ((+)-Catechin) | 154-23-4 | 290.27 | 25 MG
SDP
Catalog No. 5000389835
Encompass_Preferred
Catechin ((+)-Catechin) is an inhibitor of cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM. It demonstrates a high purity of 99.30% and is used in various research applications, including those related to cancer, neuronal cytotoxicity, and episodic memory.
  • Exhibits >95% inhibitory activity at 70 μg/mL against COX-1.
  • Induces apoptosis in MCF-7 cells in a dose- and time-dependent manner.
  • Prevents time-induced episodic memory deficits in Wistar rats.
  • Shows anti-inflammatory and antiaggregatory activities.
  • Demonstrates antiviral activity against HIV1 3B infected human H9 cells.
Catalog No. 50-003-89835 Supplier Medchemexpress LLC Supplier No. 25MGHYN0898A
Catechin ((+)-Catechin) is an inhibitor of cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM. It demonstrates a high purity of 99.30% and is used in various research applications, including those related to cancer, neuronal cytotoxicity, and episodic memory.
  • Exhibits >95% inhibitory activity at 70 μg/mL against COX-1.
  • Induces apoptosis in MCF-7 cells in a dose- and time-dependent manner.
  • Prevents time-induced episodic memory deficits in Wistar rats.
  • Shows anti-inflammatory and antiaggregatory activities.
  • Demonstrates antiviral activity against HIV1 3B infected human H9 cells.