Learn More
Medchemexpress LLC (-)-Catechin gallate | 130405-40-2 | 442.37 | 5 MG
SDP
Catalog No. 5000389142
Encompass_Preferred
(-)-Catechin gallate is a minor constituent found in green tea catechins. It functions by inhibiting the activity of COX-1 and COX-2 enzymes. Additionally, it directly interacts with DNA oligomers, inhibits gene expression of matrix metalloproteinase-9 in macrophage-differentiated HL-60 myeloid leukemia cells, inhibits adipocyte uptake of glucose by GLUT4, and inhibits the activities of various proteasomes.
  • Minor constituent in green tea catechins.
  • Inhibits COX-1 and COX-2 enzymes.
  • Directly interacts with DNA oligomers.
  • Inhibits gene expression of matrix metalloproteinase-9.
  • Inhibits adipocyte uptake of glucose by GLUT4.
  • Inhibits activities of various proteasomes.
Catalog No. 50-003-89142 Supplier Medchemexpress LLC Supplier No. HYN03565MG
(-)-Catechin gallate is a minor constituent found in green tea catechins. It functions by inhibiting the activity of COX-1 and COX-2 enzymes. Additionally, it directly interacts with DNA oligomers, inhibits gene expression of matrix metalloproteinase-9 in macrophage-differentiated HL-60 myeloid leukemia cells, inhibits adipocyte uptake of glucose by GLUT4, and inhibits the activities of various proteasomes.
  • Minor constituent in green tea catechins.
  • Inhibits COX-1 and COX-2 enzymes.
  • Directly interacts with DNA oligomers.
  • Inhibits gene expression of matrix metalloproteinase-9.
  • Inhibits adipocyte uptake of glucose by GLUT4.
  • Inhibits activities of various proteasomes.