Learn More
AdipoGen (+)-Catechin Hydrate, 225937-10-0, MFCD00150865, 1 g
SDP
Supplier:  AdipoGen AGCN20407G001
Encompass_Preferred
MF: C15H14O6 . H2O, MW: 308.284, ≥95%, Appearance: Off-white powder. Isolated from Uncaria rhynchophylla. Specific histidine decarboxylase inhibitor. Anti-inflammatory. COX-1 inhibitor. Antioxidant flavonoid. Free radical scavenger. Inhibits lipid peroxidation. Antimetastatic and anticancer compound. Antiangiogenic. Anti-osteoporotic. Antibacterial and antifungal. Neuroprotective. Mono oxidase B inhibitor. Apoptosis and cell cycle arrest inducer. JNK phosphorylation inhibitor. Antispasmodic. Insulin-mimetic.
Catalog No. 50-596-034