Learn More
Medchemexpress LLC HY-N0355 100mg Medchemexpress, (+)-Catechin hydrate CAS:225937-10-0 Purity:>98%
SDP
Catalog No. 501970257
Encompass_Preferred
Medchemexpress, HY-N0355 100mg (+)-Catechin hydrate CAS:225937-10-0 (+)-Catechin hydrate inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0257 Supplier Medchemexpress LLC Supplier No. HYN0355100MG
Medchemexpress, HY-N0355 100mg (+)-Catechin hydrate CAS:225937-10-0 (+)-Catechin hydrate inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.