Learn More
AdipoGen -Catechin hydrate
SDP
Catalog No. 502562548
Encompass_Preferred
Chemical. CAS 225937-10-0 or 88191-48-4. Formula C15H14O6 . H2O. MW 290.3 . 18.0. Isolated from Uncaria rhynchophylla. Specific histidine decarboxylase inhibitor. Anti-inflammatory. COX-1 inhibitor. Antioxidant flavonoid. Free radical scavenger. Inhibits lipid peroxidation. Antimetastatic and anticancer compound. Antiangiogenic. Anti-osteoporotic. Antibacterial and antifungal. Neuroprotective. Mono oxidase B inhibitor. Apoptosis and cell cycle arrest inducer. JNK phosphorylation inhibitor. Antispasmodic. Insulin-mimetic.
Catalog No. 50-256-2548 Supplier AdipoGen Supplier No. AGCN20407M500
Chemical. CAS 225937-10-0 or 88191-48-4. Formula C15H14O6 . H2O. MW 290.3 . 18.0. Isolated from Uncaria rhynchophylla. Specific histidine decarboxylase inhibitor. Anti-inflammatory. COX-1 inhibitor. Antioxidant flavonoid. Free radical scavenger. Inhibits lipid peroxidation. Antimetastatic and anticancer compound. Antiangiogenic. Anti-osteoporotic. Antibacterial and antifungal. Neuroprotective. Mono oxidase B inhibitor. Apoptosis and cell cycle arrest inducer. JNK phosphorylation inhibitor. Antispasmodic. Insulin-mimetic.