Learn More
Selleck Chemical LLC ()-Catechin hydrate S3974-25mg
SDP
Catalog No. 503111228
Encompass_Preferred
( )-Catechin hydrate is a plant derived polyphenolic anti-oxidants with phytotoxic properties ( )-Catechin hydrate is a frequent component of traditional herbal remedies such as Uncaria rhynchophylla ( )-Catechin hydrate inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1 4 M
Catalog No. 50-311-1228 Supplier Selleck Chemical LLC Supplier No. S397425MG
( )-Catechin hydrate is a plant derived polyphenolic anti-oxidants with phytotoxic properties ( )-Catechin hydrate is a frequent component of traditional herbal remedies such as Uncaria rhynchophylla ( )-Catechin hydrate inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1 4 M