Learn More
Apexbio Technology LLC (+)-Catechin hydrate 225937-10-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000090368
Encompass_Preferred
( )-Catechin hydrate (CAS 225937-10-0) is a plant-derived flavonoid recognized for its antioxidant properties It functions primarily as a free radical scavenger mitigating oxidative stress by neutralizing reactive oxygen species At physiological pH ( )-catechin hydrate has been shown to protect against DNA strand breaks induced by hydroxyl radicals indicating its capacity to prevent free radical-mediated cellular damage This compound is widely utilized in biomedical research to investigate cellular defense mechanisms against oxidative injury and to explore potential therapeutic strategies for conditions associated with oxidative stress
Catalog No. 50-000-90368 Supplier Apexbio Technology LLC Supplier No. N16925.1
( )-Catechin hydrate (CAS 225937-10-0) is a plant-derived flavonoid recognized for its antioxidant properties It functions primarily as a free radical scavenger mitigating oxidative stress by neutralizing reactive oxygen species At physiological pH ( )-catechin hydrate has been shown to protect against DNA strand breaks induced by hydroxyl radicals indicating its capacity to prevent free radical-mediated cellular damage This compound is widely utilized in biomedical research to investigate cellular defense mechanisms against oxidative injury and to explore potential therapeutic strategies for conditions associated with oxidative stress