Learn More
Cayman Chemical (+)-Catechin (hydrate), 225937-10-0, 10 g
SDP
Supplier:  Cayman Chemical 7094010
Encompass_Preferred
Catechin is a polyphenolic flavonoid which has been isolated from a variety of natural sources including tea leaves, grape seeds, and the wood and bark of trees such as acacia and mahogany. Catechin is a more potent antioxidant than ascorbate or α-tocopherol in certain in vitro lipid peroxidation assays.{7582} Catechin inhibits the free radical-induced oxidation of isolated LDL by AAPH.{7442} It has also been reported to act as an inhibitor of COX-1 with an IC50 value of about 80 µM.{8140} Catechin and other related procyanidin compounds have antitumor activity when tested in a two-stage mouse epidermal carcinoma model employing topical application.{7582}
Catalog No. NC0988228