Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Catechin Hydrate 97.0+%, TCI America™
SDP

Catalog No. C070510G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C070510G 10 g
C07051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C070510G Supplier TCI America Supplier No. C070510G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 154-23-4
Molecular Formula C15H14O6
Molecular Weight (g/mol) 290.271
MDL Number MFCD00075649
InChI Key PFTAWBLQPZVEMU-DZGCQCFKSA-N
Synonym (+)-3,3′C,4′C,5,7-Flavanpentol
PubChem CID 9064
ChEBI CHEBI:15600
IUPAC Name (2R,3S)-2-(3,4-dihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C=C3)O)O)O

Specifications

Melting Point 96°C
Quantity 10 g
Formula Weight 290.27
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (+)-Catechin Hydrate
RTECSNumber DJ3450000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.