Learn More
Enzo Life Sciences (+)-Catechin. monohydrate (1g). CAS: 225937-10-0
SDP
Supplier:  Enzo Life Sciences ALX385017G001
Encompass_Preferred
Antioxidant flavonoid. Free radical scavenger. Has chemopreventive and antitumor properties. Alternative name: (+)-3,3',4',5,7-Flavanpentol . H2O. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol (50mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-9047