Learn More
Medchemexpress LLC Catechin (Standard) | 154-23-4 | 290.27 | 100 MG
SDP
Catalog No. 5000416836
Encompass_Preferred
Catechin (Standard) is the analytical standard of Catechin, intended for research and analytical applications. It inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM.
  • Used as an analytical reference standard.
  • Commonly used in qualitative research experiments.
  • Commonly used in quantitative research experiments.
  • Commonly used in methodological research experiments.
  • Suitable for HPLC.
  • Suitable for GC.
  • Suitable for MS.
Catalog No. 50-004-16836 Supplier Medchemexpress LLC Supplier No. HYN0898R100MG
Catechin (Standard) is the analytical standard of Catechin, intended for research and analytical applications. It inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM.
  • Used as an analytical reference standard.
  • Commonly used in qualitative research experiments.
  • Commonly used in quantitative research experiments.
  • Commonly used in methodological research experiments.
  • Suitable for HPLC.
  • Suitable for GC.
  • Suitable for MS.