Learn More
Medchemexpress LLC Catechin (Standard) | 154-23-4 | 290.27 | 25 MG
SDP
Catalog No. 5000390141
Encompass_Preferred
Catechin (Standard) is the analytical standard of Catechin, intended for research and analytical applications. It inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM. The compound is an analytical standard grade used as a reference standard for assays and is commonly employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for Catechin
  • Intended for research and analytical applications
  • Inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Reference standard supplied assay
Catalog No. 50-003-90141 Supplier Medchemexpress LLC Supplier No. 25MGHYN0898R
Catechin (Standard) is the analytical standard of Catechin, intended for research and analytical applications. It inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM. The compound is an analytical standard grade used as a reference standard for assays and is commonly employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for Catechin
  • Intended for research and analytical applications
  • Inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Reference standard supplied assay