Learn More
Medchemexpress LLC Catechin (Standard) | 154-23-4 | 290.27 | 5 MG
SDP
Catalog No. 5000416622
Encompass_Preferred
Catechin (Standard) is the analytical standard of Catechin, intended for research and analytical applications. It inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM.
  • Analytical standard for Catechin.
  • Intended for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-16622 Supplier Medchemexpress LLC Supplier No. HYN0898R5MG
Catechin (Standard) is the analytical standard of Catechin, intended for research and analytical applications. It inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM.
  • Analytical standard for Catechin.
  • Intended for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.