Learn More
Medchemexpress LLC Catechin (Standard) | 154-23-4 | 290.27 | 50 MG
SDP
Catalog No. 5000415472
Encompass_Preferred
Catechin (Standard) is an analytical standard intended for research and analytical applications. It inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM and serves as a reference standard assay.
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments
  • Compatible with HPLC, GC, and MS techniques
Catalog No. 50-004-15472 Supplier Medchemexpress LLC Supplier No. HYN0898R50MG
Catechin (Standard) is an analytical standard intended for research and analytical applications. It inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM and serves as a reference standard assay.
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments
  • Compatible with HPLC, GC, and MS techniques