Learn More
Cayman Chemical (−)-Catechin, Purity: ≥98%, CAS: 18829-70-4, 5 mg
SDP
Catalog No. 502979521
Encompass_Preferred
A diastereoisomer of catechin having a trans 2S,3R configuration at the chiral center; a potent antioxidant that inhibits the free radical-induced oxidation of isolated LDL by AAPH
Catalog No. 50-297-9521 Supplier Cayman Chemical Supplier No. 186445 MG
A diastereoisomer of catechin having a trans 2S,3R configuration at the chiral center; a potent antioxidant that inhibits the free radical-induced oxidation of isolated LDL by AAPH