Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Catechol sulfate-d4 sodium | 00-00-0 | MFCD00015885 | 99.8% | 216.18 g/mol | C6HD4NaO5S | 5 MG
SDP

Catalog No. 5000221578
Encompass_Preferred

Catechol sulfate-d4 sodium is the deuterium-labeled (d4) sodium salt of catechol sulfate supplied for analytical and tracer applications. It is intended as an internal standard for quantitative NMR, GC-MS, and LC-MS workflows and is provided with high purity and manufacturer storage recommendations to maintain stability.

  • High purity (99.78%) for accurate quantitation.
  • Deuterium-labeled (d4) isotopologue suitable as an internal standard.
  • Sodium salt form to aid aqueous solubility.
  • Compatible with NMR, GC-MS, and LC-MS analytical methods.
  • Molecular weight ≈ 216.18 g/mol; formula C6HD4NaO5S.
  • Recommended storage: -20°C sealed; in solvent: -80°C (6 months), -20°C (1 month).
  • Available in small milligram sizes for tracer and analytical use.

Catalog No. 50-002-21578 Supplier Medchemexpress LLC Supplier No. HY144152S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Catechol sulfate-d4 sodium is the deuterium-labeled (d4) sodium salt of catechol sulfate supplied for analytical and tracer applications. It is intended as an internal standard for quantitative NMR, GC-MS, and LC-MS workflows and is provided with high purity and manufacturer storage recommendations to maintain stability.

  • High purity (99.78%) for accurate quantitation.
  • Deuterium-labeled (d4) isotopologue suitable as an internal standard.
  • Sodium salt form to aid aqueous solubility.
  • Compatible with NMR, GC-MS, and LC-MS analytical methods.
  • Molecular weight ≈ 216.18 g/mol; formula C6HD4NaO5S.
  • Recommended storage: -20°C sealed; in solvent: -80°C (6 months), -20°C (1 month).
  • Available in small milligram sizes for tracer and analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.