Learn More
Medchemexpress LLC Catechol sulfate-d4 (sodium) | 4918-96-1 | 99.8% | C6HD4NaO5S | 1 MG
SDP
Catalog No. 5000354511
Encompass_Preferred
Catechol sulfate-d4 (sodium) is the deuterium-labeled Catechol sulfate sodium. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-54511 Supplier Medchemexpress LLC Supplier No. HY144152S1MG
Add to Cart
Add to Cart
Catechol sulfate-d4 (sodium) is the deuterium-labeled Catechol sulfate sodium. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.