Learn More
Medchemexpress LLC Catechol violet | 115-41-3 | 90.0% | 250 MG
SDP
Catalog No. 5000414444
Encompass_Preferred
Catechol violet is an efficient and versatile ligand for Cu(I)-catalyzed C-S coupling reactions. It also functions as a complexometric indicator dye.
  • Efficient and versatile ligand for Cu(I)-catalyzed C-S coupling reactions.
  • Complexometric indicator dye.
Catalog No. 50-004-14444 Supplier Medchemexpress LLC Supplier No. 250MGHYD0798
Catechol violet is an efficient and versatile ligand for Cu(I)-catalyzed C-S coupling reactions. It also functions as a complexometric indicator dye.
  • Efficient and versatile ligand for Cu(I)-catalyzed C-S coupling reactions.
  • Complexometric indicator dye.