Learn More
Medchemexpress LLC Catechol violet | 115-41-3 | 90.0% | 500 MG
Supplier: Medchemexpress LLC HYD0798500MG
Catechol violet is an efficient and versatile ligand for Cu(I)-catalyzed C-S coupling reactions and a complexometric indicator dye.
- Efficient and versatile ligand for Cu(I)-catalyzed C-S coupling reactions.
- Complexometric indicator dye.