Learn More
Enzo Life Sciences beta-Catenin ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900135
Encompass_Preferred
The β-Catenin EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours that measures pan β-Catenin. Absorbance is read at 450 nm. This kit has been validated in-house with human, mouse, and rat cell lysates and correlated Western blot. The β-Catenin EIA kit detects as little 27 pg/mL β-catenin surpassing semi-quantitative Western blot analysis and provides simple liquid color-coded reagents that reduce laboratory error. Alternative Name: Cadherin-associated protein. Sensitivity: 26.8 pg/ml (range 125 - 8,000 pg/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human, Mouse, Rat. Contents: Microtiter plate, Conjugate, Antibody, Assay buffer 13, RIPA Cell lysis buffer 2, Wash buffer concentrate, Standard, TMB Substrate, Stop solution 2
Catalog No. 50-658-165